Assay Details
Target Gene Details
Entrez Gene ID: | 146225 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 2 |
Gene Aliases: |
CKLFSF2 |
Location: |
Chr.16:66579448-66588275 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM2 | NM_001199317.1 | NP_001186246.1 | ||
| NM_144673.2 | NP_653274.1 | |||
| AF479260.1 | AAN73038.1 | |||
| AK098598.1 | BAC05345.1 | |||
| AY862139.1 | AAW51946.1 | |||
| BC025354.1 | AAH25354.1 | |||
| BI830241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv123n111 | Chr.16:66568129 - 66581930 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map