Assay Details
Target Gene Details
Entrez Gene ID: | 3257 |
Gene Name: | HPS1, biogenesis of lysosomal organelles complex 3 subunit 1 |
Gene Aliases: |
BLOC3S1, HPS |
Location: |
Chr.10:98415068-98446963 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPS1 | NM_000195.4 | 20 | 3616 | NP_000186.2 |
| NM_001311345.1 | 19 | 3569 | NP_001298274.1 | |
| NM_001322476.1 | 20 | 3545 | NP_001309405.1 | |
| NM_001322477.1 | 20 | 3625 | NP_001309406.1 | |
| NM_001322478.1 | 19 | 3446 | NP_001309407.1 | |
| NM_001322479.1 | 19 | 3526 | NP_001309408.1 | |
| NM_001322480.1 | 18 | 3284 | NP_001309409.1 | |
| NM_001322481.1 | 18 | 3364 | NP_001309410.1 | |
| NM_001322482.1 | 17 | 3265 | NP_001309411.1 | |
| NM_001322483.1 | 19 | 3402 | NP_001309412.1 | |
| NM_001322484.1 | 19 | 3482 | NP_001309413.1 | |
| NM_001322485.1 | 18 | 3374 | NP_001309414.1 | |
| NM_001322487.1 | 20 | 3668 | NP_001309416.1 | |
| NM_001322489.1 | 18 | 3426 | NP_001309418.1 | |
| XM_017016170.1 | 17 | 3172 | XP_016871659.1 | |
| XM_017016171.1 | 18 | 3379 | XP_016871660.1 | |
| XM_017016172.1 | 17 | 3190 | XP_016871661.1 | |
| BF447504.1 | 1 | 130 | ||
| U65676.1 | 20 | 3560 | AAB17869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524427 | Chr.10:98408615 - 98452617 on Build GRCh38 | Loss |
|
| nsv515723 | Chr.10:98388419 - 98435072 on Build GRCh38 | Loss |
|
| nsv831955 | Chr.10:98402137 - 98592058 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map