Assay Details
Target Gene Details
Entrez Gene ID: | 904 |
Gene Name: | cyclin T1 |
Gene Aliases: |
CCNT, CYCT1, HIVE1 |
Location: |
Chr.12:48688458-48716998 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNT1 | NM_001240.3 | 9 | 2172 | NP_001231.2 |
| NM_001277842.1 | 8 | 2008 | NP_001264771.1 | |
| XM_017020197.1 | 8 | 1683 | XP_016875686.1 | |
| AF045161.1 | 9 | 1849 | AAC39638.1 | |
| AF048730.1 | 9 | 1893 | AAC39664.1 | |
| AK290990.1 | 9 | 2072 | ||
| EF688064.1 | 8 | 1685 |
More Information
Set Membership: |
|

Genomic Map