Assay Details
Target Gene Details
Entrez Gene ID: | 9361 |
Gene Name: | lon peptidase 1, mitochondrial |
Gene Aliases: |
CODASS, LON, LONP, LonHS, PIM1, PRSS15, hLON |
Location: |
Chr.19:5691834-5720452 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LONP1 | NM_001276479.1 | NP_001263408.1 | ||
| NM_001276480.1 | NP_001263409.1 | |||
| NM_004793.3 | NP_004784.2 | |||
| NR_076392.1 | ||||
| AK056366.1 | ||||
| AK096433.1 | ||||
| AK096626.1 | BAC04829.1 | |||
| AK127867.1 | ||||
| AK298531.1 | ||||
| AK309720.1 | ||||
| AL096720.1 | ||||
| BC000235.1 | AAH00235.1 | |||
| BC109218.1 | AAI09219.1 | |||
| U02389.1 | AAA61616.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953952 | Chr.19:5645090 - 5708389 on Build GRCh38 | Deletion |
|
| nsv1062851 | Chr.19:5531540 - 5893590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map