Assay Details
Target Gene Details
Entrez Gene ID: | 10309 |
Gene Name: | cyclin O |
Gene Aliases: |
CCNU, CILD29, UDG2 |
Location: |
Chr.5:55231152-55233717 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNO | NM_021147.4 | 3 | 1284 | NP_066970.3 |
| NR_125346.1 | 3 | 1660 | ||
| NR_125347.1 | 3 | 1289 | ||
| NR_125348.1 | 2 | 1154 | ||
| AK026075.1 | 3 | 1247 | BAB15351.1 | |
| BC004877.2 | 3 | 1540 | ||
| X52486.1 | 2 | 1054 | CAA36728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830302 | Chr.5:55107550 - 55259351 on Build GRCh38 | Gain |
|
| nsv1027090 | Chr.5:55179438 - 55375973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map