Assay Details
Target Gene Details
Entrez Gene ID: | 93343 |
Gene Name: | multivesicular body subunit 12A |
Gene Aliases: |
CFBP, FAM125A |
Location: |
Chr.19:17405686-17425339 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MVB12A | NM_001304547.1 | NP_001291476.1 | ||
| NM_138401.3 | NP_612410.1 | |||
| AK057057.1 | ||||
| AK289487.1 | ||||
| BC007883.2 | AAH07883.2 | |||
| BC011840.2 | AAH11840.1 | |||
| BG684013.1 | ||||
| BY996463.1 | ||||
| CD672349.1 | ||||
| DR006745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833768 | Chr.19:17338840 - 17526738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map