Assay Details
Target Gene Details
Entrez Gene ID: | 51651 |
Gene Name: | peptidyl-tRNA hydrolase 2 |
Gene Aliases: |
BIT1, CFAP37, CGI-147, IMNEPD, PTH, PTH 2, PTH2 |
Location: |
Chr.17:59697306-59707598 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRH2 | NM_001015509.2 | NP_001015509.1 | ||
| NM_016077.4 | NP_057161.1 | |||
| XM_011524887.1 | XP_011523189.1 | |||
| AF151905.1 | AAD34142.1 | |||
| AK057033.1 | ||||
| AK098219.1 | 1 | 1574 | ||
| AL137322.1 | CAB70696.1 | |||
| BC006807.1 | AAH06807.1 | |||
| BX647177.1 | 1 | 3505 | ||
| DA092491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833501 | Chr.17:59627926 - 59779793 on Build GRCh38 | Loss |
|
| esv32651 | Chr.17:59697677 - 59701071 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map