Assay Details
Target Gene Details
Entrez Gene ID: | 3587 |
Gene Name: | interleukin 10 receptor subunit alpha |
Gene Aliases: |
CD210, CD210a, CDW210A, HIL-10R, IL-10R1, IL10R |
Location: |
Chr.11:117986391-118001483 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL10RA | NM_001558.3 | 7 | 2029 | NP_001549.2 |
| NR_026691.1 | 8 | 2159 | ||
| AB209626.1 | 6 | 3530 | BAD92863.1 | |
| AK291645.1 | 7 | 2029 | ||
| AK301484.1 | ||||
| AK316146.1 | ||||
| AK316342.1 | 8 | 2233 | ||
| BC028082.1 | 7 | 2006 | AAH28082.1 | |
| U00672.1 | 7 | 2013 | AAA17896.1 |
More Information
Set Membership: |
|

Genomic Map