Assay Details
Target Gene Details
Entrez Gene ID: | 25865 |
Gene Name: | protein kinase D2 |
Gene Aliases: |
HSPC187, PKD2, nPKC-D2 |
Location: |
Chr.19:46674316-46717127 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKD2 | NM_001079880.1 | 6 | 1251 | NP_001073349.1 |
| NM_001079881.1 | 6 | 1115 | NP_001073350.1 | |
| NM_001079882.1 | 4 | 852 | NP_001073351.1 | |
| NM_016457.4 | 5 | 1530 | NP_057541.2 | |
| XM_005258716.2 | 4 | 508 | XP_005258773.2 | |
| AF309082.1 | 5 | 812 | AAK01149.1 | |
| AK074673.1 | 4 | 491 | BAC11127.1 | |
| AK075266.1 | 4 | 601 | BAC11508.1 | |
| AK092896.1 | 5 | 903 | ||
| AK095884.1 | 6 | 794 | ||
| AK300339.1 | 6 | 1065 | ||
| AK303629.1 | 5 | 715 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map