Assay Details
Target Gene Details
Entrez Gene ID: | 8399 |
Gene Name: | phospholipase A2 group X |
Gene Aliases: |
GXPLA2, GXSPLA2, SPLA2, sPLA2-X |
Location: |
Chr.16:14672548-14694663 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G10 | NM_003561.2 | NP_003552.1 | ||
| NR_133651.1 | ||||
| BC069539.1 | AAH69539.1 | |||
| BC106731.1 | AAI06732.1 | |||
| BC106732.1 | AAI06733.1 | |||
| BC111804.1 | ||||
| BM739473.1 | ||||
| CR456885.1 | CAG33166.1 | |||
| U95301.1 | AAB64410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833144 | Chr.16:14587299 - 14813650 on Build GRCh38 | Gain |
|
| dgv2738n100 | Chr.16:14617525 - 15031584 on Build GRCh38 | Gain |
|
| esv2758632 | Chr.16:14528198 - 15272693 on Build GRCh38 | Gain+Loss |
|
| nsv1740 | Chr.16:14654306 - 14769738 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map