Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GORASP1 | NM_001278789.1 | 7 | 1661 | NP_001265718.1 |
| NM_001278790.1 | 6 | 1481 | NP_001265719.1 | |
| NM_031899.3 | 9 | 2409 | NP_114105.1 | |
| NR_103866.1 | 9 | 1905 | ||
| NR_103867.1 | 9 | 1897 | ||
| XM_006713301.3 | 9 | 2509 | XP_006713364.1 | |
| XM_011534020.2 | 9 | 2419 | XP_011532322.1 | |
| XM_011534021.2 | 8 | 2329 | XP_011532323.1 | |
| XM_017007050.1 | 9 | 1595 | XP_016862539.1 | |
| XM_017007051.1 | 8 | 1944 | XP_016862540.1 | |
| XM_017007052.1 | 8 | 1540 | XP_016862541.1 | |
| AJ409349.1 | 9 | 1685 | CAC35160.1 | |
| AK001574.1 | 8 | 1927 | ||
| AK057043.1 | 6 | 1219 | ||
| AK091168.1 | 9 | 1833 | BAC03598.1 | |
| AK124755.1 | 9 | 2409 | ||
| BC008928.2 | 9 | 1626 | AAH08928.1 | |
| BC075854.1 | 9 | 1695 | AAH75854.1 | |
| BC103826.1 | 9 | 1627 | AAI03827.1 | |
| BC103827.1 | 9 | 1627 | AAI03828.1 | |
| BM470731.1 |
More Information
Set Membership: |
|

Genomic Map