Assay Details
Target Gene Details
Entrez Gene ID: | 6446 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
SGK |
Location: |
Chr.6:134169246-134318112 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGK1 | NM_001143676.1 | NP_001137148.1 | ||
| NM_001143677.1 | NP_001137149.1 | |||
| NM_001143678.1 | NP_001137150.1 | |||
| NM_001291995.1 | NP_001278924.1 | |||
| NM_005627.3 | NP_005618.2 | |||
| XM_011536071.1 | XP_011534373.1 | |||
| AF153609.1 | AAD41091.1 | |||
| AK055077.1 | ||||
| AK098509.1 | ||||
| AK293383.1 | ||||
| AK295422.1 | ||||
| AK298688.1 | ||||
| BC001263.1 | AAH01263.1 | |||
| BX649005.1 | 2 | 1974 | CAH10381.1 | |
| EU518415.1 | ||||
| FM205707.1 | ||||
| FM205708.1 | ||||
| FM205709.1 | ||||
| FM205710.1 | ||||
| Y10032.1 | CAA71138.1 |
More Information
Set Membership: |
|

Genomic Map