Assay Details
Target Gene Details
Entrez Gene ID: | 112744 |
Gene Name: | interleukin 17F |
Gene Aliases: |
CANDF6, IL-17F, ML-1, ML1 |
Location: |
Chr.6:52236681-52245689 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL17F | NM_052872.3 | 2 | 204 | NP_443104.1 |
| XM_011514276.1 | 3 | 270 | XP_011512578.1 | |
| AF332389.1 | 1 | 317 | AAL14427.1 | |
| AF384857.1 | 2 | 133 | AAK83350.1 | |
| BC070124.1 | 2 | 204 | AAH70124.1 | |
| JF305974.1 | 2 | 133 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830662 | Chr.6:52064873 - 52264478 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map