Assay Details
Target Gene Details
Entrez Gene ID: | 166968 |
Gene Name: | MIER family member 3 |
Gene Aliases: |
- |
Location: |
Chr.5:56919602-56952130 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIER3 | NM_001297598.1 | 13 | 2134 | NP_001284527.1 |
| NM_001297599.1 | 13 | 2119 | NP_001284528.1 | |
| NM_152622.4 | 13 | 2116 | NP_689835.3 | |
| XM_011543216.1 | 13 | 2154 | XP_011541518.1 | |
| XM_011543217.1 | 13 | 2151 | XP_011541519.1 | |
| XM_017009149.1 | 14 | 2155 | XP_016864638.1 | |
| AK002183.1 | ||||
| AK093273.1 | 11 | 2046 | BAC04117.1 | |
| BC041348.2 | 13 | 2113 | AAH41348.2 | |
| BX537798.1 | 13 | 2104 | CAD97846.1 | |
| BX640907.1 | 13 | 2122 | CAE45951.2 | |
| BX647359.1 | 13 | 2110 | ||
| BX648294.1 | 13 | 2111 | CAI56710.1 | |
| CR749246.1 | 14 | 2154 | CAH18102.1 | |
| CR749805.1 | 14 | 2154 | CAH18665.1 |
Target Gene Details
Entrez Gene ID: | 133383 |
Gene Name: | SET domain containing 9 |
Gene Aliases: |
C5orf35 |
Location: |
Chr.5:56909260-56928900 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD9 | NM_001171990.2 | NP_001165461.1 | ||
| NR_136555.1 | ||||
| AK309152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598210 | Chr.5:56881616 - 56958658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map