Assay Details
Target Gene Details
Entrez Gene ID: | 56994 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
CPT, CPT1 |
Location: |
Chr.12:101697639-101752038 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHPT1 | NM_020244.2 | 2 | 574 | NP_064629.2 |
| XM_011538574.1 | 2 | 541 | XP_011536876.1 | |
| XM_011538575.1 | 1 | 78 | XP_011536877.1 | |
| AF047431.1 | 2 | 145 | AAD44019.1 | |
| AF111803.1 | AAL39005.1 | |||
| AF195623.1 | 2 | 521 | AAF87947.1 | |
| AF195624.1 | 2 | 521 | AAF87948.1 | |
| AK075211.1 | 2 | 541 | ||
| AK093067.1 | 3 | 433 | ||
| AK226127.1 | 2 | 248 | ||
| BC020819.1 | 2 | 528 | AAH20819.1 | |
| BC050429.1 | 2 | 567 | AAH50429.1 | |
| CD300296.1 | 2 | 600 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832497 | Chr.12:101667198 - 101861862 on Build GRCh38 | Gain |
|
| dgv2836n54 | Chr.12:101711350 - 101716557 on Build GRCh38 | Loss |
|
| dgv2838n54 | Chr.12:101711851 - 101723811 on Build GRCh38 | Loss |
|
| nsv560014 | Chr.12:101711211 - 101714567 on Build GRCh38 | Loss |
|
| nsv859 | Chr.12:101696419 - 101720084 on Build GRCh38 | Deletion |
|
| nsv560006 | Chr.12:101703234 - 101714567 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map