Assay Details
Target Gene Details
Entrez Gene ID: | 339479 |
Gene Name: | BMP/retinoic acid inducible neural specific 3 |
Gene Aliases: |
DBCCR1L, DBCCR1L1, FAM5C |
Location: |
Chr.1:190097662-190477997 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRINP3 | NM_001317188.1 | 7 | 1536 | NP_001304117.1 |
| NM_199051.2 | 8 | 1931 | NP_950252.1 | |
| XM_011509472.2 | 9 | 1933 | XP_011507774.1 | |
| XM_011509475.2 | 8 | 1671 | XP_011507777.1 | |
| XM_011509476.2 | 7 | 1749 | XP_011507778.1 | |
| XM_017001125.1 | 10 | 2042 | XP_016856614.1 | |
| XM_017001126.1 | 9 | 1907 | XP_016856615.1 | |
| XM_017001127.1 | 9 | 2071 | XP_016856616.1 | |
| XM_017001128.1 | 9 | 3159 | XP_016856617.1 | |
| XM_017001129.1 | 7 | 1740 | XP_016856618.1 | |
| XM_017001130.1 | 8 | 1716 | XP_016856619.1 | |
| XM_017001131.1 | 9 | 2021 | XP_016856620.1 | |
| AB111893.1 | 8 | 1678 | BAD04066.1 | |
| AK123024.1 | 8 | 1621 | ||
| AK123637.1 | 8 | 1676 | ||
| AK294365.1 | 7 | 1338 | ||
| BC105052.1 | 7 | 1502 | AAI05053.1 | |
| BC105054.1 | 7 | 1502 | AAI05055.1 | |
| U67037.1 | 2 | 477 | AAD09521.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762036 | Chr.1:190073576 - 190122891 on Build GRCh38 | Gain |
|
| dgv693n54 | Chr.1:190067010 - 190110606 on Build GRCh38 | Loss |
|
| nsv832104 | Chr.1:190021018 - 190217578 on Build GRCh38 | Gain |
|
| dgv694n54 | Chr.1:190094081 - 190147232 on Build GRCh38 | Loss |
|
| nsv548532 | Chr.1:189789642 - 190402541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map