Assay Details
Target Gene Details
Entrez Gene ID: | 9945 |
Gene Name: | glutamine-fructose-6-phosphate transaminase 2 |
Gene Aliases: |
GFAT, GFAT 2, GFAT2 |
Location: |
Chr.5:180300690-180353387 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFPT2 | NM_005110.3 | 19 | 2855 | NP_005101.1 |
| XM_006714942.2 | 18 | 2642 | XP_006715005.1 | |
| XM_017010100.1 | 18 | 2740 | XP_016865589.1 | |
| AB016789.1 | 19 | 2784 | BAA74731.1 | |
| AK001242.1 | 9 | 1737 | ||
| AK022507.1 | 19 | 2795 | ||
| BC000012.2 | 19 | 2793 | AAH00012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10787 | Chr.5:180290724 - 180305388 on Build GRCh38 | Gain |
|
| nsv1033706 | Chr.5:179975769 - 180379356 on Build GRCh38 | Gain |
|
| esv2759398 | Chr.5:180107477 - 180302944 on Build GRCh38 | Gain |
|
| nsv1026417 | Chr.5:180294376 - 180864360 on Build GRCh38 | Gain |
|
| nsv830563 | Chr.5:180290693 - 180471609 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79423981] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map