Assay Details
Target Gene Details
Entrez Gene ID: | 643 |
Gene Name: | C-X-C motif chemokine receptor 5 |
Gene Aliases: |
BLR1, CD185, MDR15 |
Location: |
Chr.11:118883766-118896271 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR5 | NM_001716.4 | 2 | 1285 | NP_001707.1 |
| NM_032966.2 | 1 | 1262 | NP_116743.1 | |
| AK291512.1 | 2 | 1285 | ||
| BC110352.1 | 2 | 1199 | AAI10353.1 | |
| CO958128.1 | ||||
| X68149.1 | 2 | 1193 | CAA48252.1 | |
| X68829.1 | 1 | 1262 | CAA48723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952743 | Chr.11:118892392 - 118923091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map