Assay Details
Target Gene Details
Entrez Gene ID: | 23155 |
Gene Name: | chloride channel CLIC like 1 |
Gene Aliases: |
MCLC |
Location: |
Chr.1:108929508-108963499 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCC1 | NM_001048210.2 | NP_001041675.1 | ||
| NM_001278202.1 | NP_001265131.1 | |||
| NM_001278203.1 | NP_001265132.1 | |||
| NM_015127.4 | NP_055942.1 | |||
| AB018304.2 | BAA34481.2 | |||
| AB052915.1 | BAB59018.1 | |||
| AB052916.1 | BAB79261.1 | |||
| AB052917.1 | BAB79262.1 | |||
| AB052918.1 | BAB79263.1 | |||
| AK090563.1 | ||||
| BC002939.2 | AAH02939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv15n21 | Chr.1:108873603 - 108941145 on Build GRCh38 | Gain |
|
| nsv526049 | Chr.1:108930432 - 108941353 on Build GRCh38 | Loss |
|
| dgv251n100 | Chr.1:108930151 - 108955511 on Build GRCh38 | Loss |
|
| nsv831026 | Chr.1:108924711 - 109119614 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map