Assay Details
Target Gene Details
Entrez Gene ID: | 1154 |
Gene Name: | cytokine inducible SH2 containing protein |
Gene Aliases: |
BACTS2, CIS, CIS-1, G18, SOCS |
Location: |
Chr.3:50606454-50611831 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISH | NM_013324.5 | 4 | 1223 | NP_037456.5 |
| NM_145071.2 | 3 | 1066 | NP_659508.1 | |
| XM_011533329.2 | 2 | 3888 | XP_011531631.1 | |
| AF035947.1 | 2 | 4034 | AAF97410.1 | |
| AF132297.2 | 4 | 1164 | AAD28471.2 | |
| BC031590.1 | 3 | 998 | AAH31590.1 | |
| BC064354.1 | 3 | 903 | AAH64354.1 | |
| D83532.1 | 3 | 996 | BAA92328.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584502 | Chr.3:50327055 - 50622310 on Build GRCh38 | Gain |
|
| nsv508919 | Chr.3:50596756 - 50676338 on Build GRCh38 | Insertion |
|
| nsv590273 | Chr.3:50536782 - 50640720 on Build GRCh38 | Loss |
|
| nsv482083 | Chr.3:50606455 - 50611831 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map