Assay Details
Target Gene Details
Entrez Gene ID: | 10466 |
Gene Name: | component of oligomeric golgi complex 5 |
Gene Aliases: |
CDG2I, GOLTC1, GTC90 |
Location: |
Chr.7:107201743-107564514 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG5 | NM_006348.3 | 22 | 3991 | NP_006339.3 |
| NM_181733.2 | 21 | 3928 | NP_859422.2 | |
| AK022835.1 | 13 | 2367 | ||
| AU117533.1 | 1 | 460 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831087 | Chr.7:107033020 - 107232015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map