Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOPEY1 | NM_001199942.1 | 32 | 6539 | NP_001186871.1 |
| NM_015018.3 | 32 | 6566 | NP_055833.2 | |
| XM_011535619.2 | 31 | 6531 | XP_011533921.1 | |
| XM_017010559.1 | 32 | 6561 | XP_016866048.1 | |
| XM_017010560.1 | 31 | 6489 | XP_016866049.1 | |
| XM_017010561.1 | 32 | 6513 | XP_016866050.1 | |
| XM_017010562.1 | 31 | 6462 | XP_016866051.1 | |
| XM_017010563.1 | 32 | 6540 | XP_016866052.1 | |
| XM_017010564.1 | 32 | 6540 | XP_016866053.1 | |
| XM_017010565.1 | 32 | 6534 | XP_016866054.1 | |
| XM_017010566.1 | 32 | 6534 | XP_016866055.1 | |
| XM_017010567.1 | 31 | 6462 | XP_016866056.1 | |
| XM_017010568.1 | 31 | 6531 | XP_016866057.1 | |
| XM_017010569.1 | 30 | 6448 | XP_016866058.1 | |
| XM_017010570.1 | 31 | 6462 | XP_016866059.1 | |
| XM_017010571.1 | 31 | 6456 | XP_016866060.1 | |
| XM_017010572.1 | XP_016866061.1 | |||
| XM_017010573.1 | 32 | 6561 | XP_016866062.1 | |
| XM_017010574.1 | 32 | 6561 | XP_016866063.1 | |
| AB029040.2 | 15 | 3587 | BAA83069.2 | |
| AK027030.1 | 12 | 2333 | BAB15631.1 | |
| AK092929.1 | ||||
| AL050080.1 | 4 | 295 | CAB43259.1 | |
| BC048342.1 | 6 | 579 | AAH48342.1 | |
| BC144544.1 | 32 | 6437 | ||
| BC150619.1 | 32 | 6437 | ||
| BX648667.1 | 32 | 6564 |
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGM3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map