Assay Details
Target Gene Details
Entrez Gene ID: | 3358 |
Gene Name: | 5-hydroxytryptamine receptor 2C |
Gene Aliases: |
5-HT1C, 5-HT2C, 5-HTR2C, 5HTR2C, HTR1C |
Location: |
Chr.X:114584078-114910061 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR2C | NM_000868.3 | 1 | 83 | NP_000859.1 |
| NM_001256760.2 | 1 | 83 | NP_001243689.1 | |
| NM_001256761.2 | 1 | 83 | NP_001243690.1 | |
| AK295753.1 | 1 | 75 | ||
| M81778.1 | 1 | 51 | AAA60317.1 | |
| U49516.1 | 1 | 83 | AAB40898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527347 | Chr.X:114537428 - 114983201 on Build GRCh38 | Gain |
|
| esv33994 | Chr.X:113685854 - 115738426 on Build GRCh38 | Gain+Loss |
|
| dgv523n21 | Chr.X:113183439 - 114588429 on Build GRCh38 | Loss |
|
| dgv524n21 | Chr.X:114067803 - 114619628 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map