Assay Details
Target Gene Details
Entrez Gene ID: | 10553 |
Gene Name: | HIV-1 Tat interactive protein 2 |
Gene Aliases: |
CC3, SDR44U1, TIP30 |
Location: |
Chr.11:20363685-20383783 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTATIP2 | NM_001098520.1 | NP_001091990.1 | ||
| NM_001098521.1 | NP_001091991.1 | |||
| NM_001098522.1 | NP_001091992.1 | |||
| NM_006410.4 | NP_006401.3 | |||
| AF039103.1 | AAC39694.1 | |||
| AK222969.1 | BAD96689.1 | |||
| AK223010.1 | BAD96730.1 | |||
| AK292092.1 | ||||
| BC002439.2 | ||||
| BC015358.2 | AAH15358.1 | |||
| U69161.1 | AAB84360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553678 | Chr.11:20097752 - 20718575 on Build GRCh38 | Gain |
|
| nsv832083 | Chr.11:20318953 - 20490151 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map