Assay Details
Target Gene Details
Entrez Gene ID: | 158584 |
Gene Name: | fatty acid amide hydrolase 2 |
Gene Aliases: |
AMDD |
Location: |
Chr.X:57121861-57489196 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAH2 | XM_011530767.2 | XP_011529069.1 |
Target Gene Details
Entrez Gene ID: | 54466 |
Gene Name: | spindlin family member 2A |
Gene Aliases: |
DXF34, SPIN2, TDRD25, dJ323P24.1 |
Location: |
Chr.X:57134259-57138688 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPIN2A | NM_019003.3 | 1 | 121 | NP_061876.3 |
| XM_005262019.3 | 1 | 1184 | XP_005262076.1 | |
| XM_017029598.1 | 1 | 1184 | XP_016885087.1 | |
| BC071694.1 | 1 | 121 | AAH71694.1 |
More Information
Set Membership: |
|

Genomic Map