Assay Details
Target Gene Details
Entrez Gene ID: | 375307 |
Gene Name: | ciliogenesis associated TTC17 interacting protein |
Gene Aliases: |
C2orf62 |
Location: |
Chr.2:218356757-218368393 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CATIP | NM_001320865.1 | 6 | 606 | NP_001307794.1 |
| NM_198559.1 | 6 | 573 | NP_940961.1 | |
| XM_005246539.4 | 5 | 570 | XP_005246596.1 | |
| XM_005246541.4 | 4 | 369 | XP_005246598.1 | |
| XM_011511148.2 | XP_011509450.1 | |||
| XM_011511149.2 | 4 | 368 | XP_011509451.1 | |
| XM_011511150.2 | 4 | 334 | XP_011509452.1 | |
| XM_017004053.1 | 4 | 769 | XP_016859542.1 | |
| BC052750.1 | 6 | 573 | AAH52750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519829 | Chr.2:218286495 - 218403058 on Build GRCh38 | Loss |
|
| esv2763102 | Chr.2:218353805 - 218374015 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map