Assay Details
Target Gene Details
Entrez Gene ID: | 55187 |
Gene Name: | vacuolar protein sorting 13 homolog D |
Gene Aliases: |
- |
Location: |
Chr.1:12230039-12512047 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS13D | NM_015378.3 | NP_056193.2 | ||
| NM_018156.3 | NP_060626.2 | |||
| AJ608774.1 | CAE75586.1 | |||
| AJ608775.1 | CAE75587.1 | |||
| AL162331.1 | CAB82724.1 | |||
| HY121734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471622 | Chr.1:12155207 - 12371588 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map