Assay Details
Target Gene Details
Entrez Gene ID: | 57590 |
Gene Name: | WD repeat and FYVE domain containing 1 |
Gene Aliases: |
FENS-1, WDF1, ZFYVE17 |
Location: |
Chr.2:223875343-223945387 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY1 | NM_020830.4 | 12 | 1733 | NP_065881.1 |
| AB037856.1 | 12 | 1646 | ||
| AJ310568.1 | 12 | 1720 | CAC83947.1 | |
| AK022888.1 | 12 | 1659 | BAB14294.1 | |
| AK023415.1 | 11 | 1527 | BAB14569.1 | |
| BC040525.1 | 12 | 1645 | AAH40525.1 | |
| BC065934.1 | 12 | 1677 | AAH65934.1 | |
| N27710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834555 | Chr.2:223728131 - 223887704 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map