Assay Details
Target Gene Details
Entrez Gene ID: | 23582 |
Gene Name: | cyclin D1 binding protein 1 |
Gene Aliases: |
DIP1, GCIP, HHM |
Location: |
Chr.15:43185118-43197177 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNDBP1 | NM_012142.4 | 1 | 233 | NP_036274.3 |
| NR_027513.2 | 1 | 233 | ||
| NR_027514.2 | 1 | 233 | ||
| NR_045998.1 | 1 | 233 | ||
| NR_045999.1 | 1 | 233 | ||
| XM_006720448.1 | 1 | 295 | XP_006720511.1 | |
| AF082569.1 | 1 | 89 | AAD11777.1 | |
| AF087852.1 | 1 | 81 | AAP97163.1 | |
| AF132034.1 | 1 | 48 | AAF77613.1 | |
| AF246144.1 | 1 | 51 | AAF67182.1 | |
| AK075146.1 | 1 | 62 | BAC11433.1 | |
| AK225143.1 | 1 | 48 | ||
| AK225645.1 | 1 | 62 | ||
| AK290665.1 | 1 | 62 | ||
| AK290707.1 | 1 | 95 | ||
| AK295017.1 | 1 | 62 | ||
| AK308817.1 | 1 | 383 | ||
| AL552334.3 | 1 | 174 | ||
| BC009689.1 | 1 | 76 | AAH09689.1 | |
| BQ439439.1 | 1 | 47 | ||
| CR450331.1 | CAG29327.1 | |||
| DA210890.1 | 1 | 383 | ||
| DA506028.1 | 1 | 222 | ||
| DB492361.1 | 1 | 233 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525461 | Chr.15:42770350 - 43189929 on Build GRCh38 | Loss |
|
| nsv569241 | Chr.15:42908911 - 43230724 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map