Assay Details
Target Gene Details
Entrez Gene ID: | 10205 |
Gene Name: | myelin protein zero like 2 |
Gene Aliases: |
EVA, EVA1 |
Location: |
Chr.11:118253416-118264536 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZL2 | NM_005797.3 | 6 | 2743 | NP_005788.1 |
| AF275945.1 | 6 | 2489 | AAF87240.1 | |
| AF304447.1 | 6 | 2484 | AAG23183.1 | |
| AW073724.1 | 1 | 129 |
More Information
Set Membership: |
|

Genomic Map