Assay Details
Target Gene Details
Entrez Gene ID: | 9141 |
Gene Name: | programmed cell death 5 |
Gene Aliases: |
TFAR19 |
Location: |
Chr.19:32581161-32587452 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDCD5 | NM_004708.3 | NP_004699.1 | ||
| XM_005259392.4 | XP_005259449.1 | |||
| AB209040.1 | BAD92277.1 | |||
| AF014955.1 | AAD11579.1 | |||
| AK293486.1 | ||||
| BC015519.2 | AAH15519.1 | |||
| BM696336.1 | ||||
| BT006694.1 | AAP35340.1 | |||
| CR456934.1 | CAG33215.1 | |||
| DN915905.1 | ||||
| DQ208400.1 | ABA60375.1 | |||
| N40187.1 |
More Information
Set Membership: |
|

Genomic Map