Assay Details
Target Gene Details
Entrez Gene ID: | 114 |
Gene Name: | adenylate cyclase 8 |
Gene Aliases: |
AC8, ADCY3, HBAC1 |
Location: |
Chr.8:130780300-131041604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY8 | NM_001115.2 | NP_001106.1 | ||
| XM_005250769.3 | XP_005250826.1 | |||
| XM_006716501.3 | XP_006716564.1 | |||
| XM_017013006.1 | XP_016868495.1 | |||
| DQ104739.1 | ||||
| GQ472223.1 | ||||
| M83533.1 | AAA35523.2 | |||
| Z35309.1 | CAA84552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016494 | Chr.8:130133708 - 131222639 on Build GRCh38 | Gain |
|
| nsv612294 | Chr.8:130632833 - 130911131 on Build GRCh38 | Gain |
|
| nsv831462 | Chr.8:130691873 - 130883547 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map