Assay Details
Target Gene Details
Entrez Gene ID: | 119504 |
Gene Name: | anaphase promoting complex subunit 16 |
Gene Aliases: |
APC16, C10orf104, CENP-27, MSAG, bA570G20.3 |
Location: |
Chr.10:72216000-72235860 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC16 | NM_001242546.1 | 5 | 2368 | NP_001229475.1 |
| NM_001242547.1 | 4 | 1976 | NP_001229476.1 | |
| NM_001242548.1 | 3 | 1901 | NP_001229477.1 | |
| NM_173473.3 | 4 | 2042 | NP_775744.1 | |
| NR_038391.1 | 3 | 1807 | ||
| NR_038392.1 | 3 | 1873 | ||
| AK023981.1 | 1 | 925 | ||
| BF975718.1 |
More Information
Set Membership: |
|

Genomic Map