Assay Details
Target Gene Details
Entrez Gene ID: | 55033 |
Gene Name: | FK506 binding protein 14 |
Gene Aliases: |
EDSKMH, FKBP22, IPBP12 |
Location: |
Chr.7:30010583-30026801 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP14 | NM_017946.3 | 4 | 1261 | NP_060416.1 |
| NR_046478.1 | 5 | 1353 | ||
| NR_046479.1 | 3 | 1109 | ||
| AK000738.1 | 4 | 1113 | BAA91351.1 | |
| AK309552.1 | 3 | 970 | ||
| AY358643.1 | 4 | 1141 | AAQ89006.1 | |
| BC005206.1 | AAH05206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019527 | Chr.7:29722658 - 30413302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map