Assay Details
Target Gene Details
Entrez Gene ID: | 5141 |
Gene Name: | phosphodiesterase 4A |
Gene Aliases: |
DPDE2, PDE4, PDE46 |
Location: |
Chr.19:10416773-10469631 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE4A | NM_001111307.1 | NP_001104777.1 | ||
| NM_001111308.1 | NP_001104778.1 | |||
| NM_001111309.1 | NP_001104779.1 | |||
| NM_001243121.1 | NP_001230050.1 | |||
| NM_006202.2 | NP_006193.1 | |||
| XM_011528054.1 | XP_011526356.1 | |||
| XM_011528055.2 | XP_011526357.1 | |||
| XM_017026865.1 | XP_016882354.1 | |||
| AF073745.2 | AAD34217.2 | |||
| AY593872.1 | AAT00628.1 | |||
| AY618547.1 | AAU82096.1 | |||
| BC019864.1 | AAH19864.1 | |||
| BC038234.1 | AAH38234.1 | |||
| L20965.1 | AAA03588.1 | |||
| M37744.1 | AAA69697.1 | |||
| S75213.1 | AAB33798.1 | |||
| U18087.1 | AAC50458.1 | |||
| U68532.1 | AAC63832.1 | |||
| U97584.1 | AAC25679.1 |
More Information
Set Membership: |
|

Genomic Map