Assay Details
Target Gene Details
Entrez Gene ID: | 50616 |
Gene Name: | interleukin 22 |
Gene Aliases: |
IL-21, IL-22, IL-D110, IL-TIF, ILTIF, TIFIL-23, TIFa, zcyto18 |
Location: |
Chr.12:68248245-68253501 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL22 | NM_020525.4 | 5 | 759 | NP_065386.1 |
| AF279437.1 | 5 | 763 | AAG22064.1 | |
| AY358890.1 | 5 | 763 | AAQ89249.1 | |
| BC070261.1 | 6 | 774 | AAH70261.1 |
More Information
Set Membership: |
|

Genomic Map