Assay Details
Target Gene Details
Entrez Gene ID: | 55709 |
Gene Name: | kelch repeat and BTB domain containing 4 |
Gene Aliases: |
BKLHD4, HSPC252 |
Location: |
Chr.11:47572197-47579015 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KBTBD4 | NM_001318716.1 | 4 | 1808 | NP_001305645.1 |
| NM_001318717.1 | 4 | 1729 | NP_001305646.1 | |
| NM_001318718.1 | 4 | 1709 | NP_001305647.1 | |
| NM_001318719.1 | 4 | 1687 | NP_001305648.1 | |
| NM_001318720.1 | 4 | 1681 | NP_001305649.1 | |
| NM_001318721.1 | 4 | 1656 | NP_001305650.1 | |
| NM_001318722.1 | 4 | 1677 | NP_001305651.1 | |
| NM_001318723.1 | 4 | 1761 | NP_001305652.1 | |
| NM_001318724.1 | 4 | 1761 | NP_001305653.1 | |
| NM_001318725.1 | 4 | 1664 | NP_001305654.1 | |
| NM_016506.6 | 4 | 1765 | NP_057590.3 | |
| NM_018095.5 | 4 | 1660 | NP_060565.4 | |
| XM_017018009.1 | 4 | 1644 | XP_016873498.1 | |
| AK001312.1 | 4 | 1614 | BAA91616.1 | |
| AK001749.1 | 4 | 1701 | BAA91880.1 | |
| AK091607.1 | 3 | 2338 | ||
| AK222550.1 | 4 | 1611 | BAD96270.1 | |
| AK222985.1 | ||||
| AK225062.1 | 4 | 1611 | ||
| AL713735.1 | 3 | 1590 | CAD28521.1 | |
| BC002736.2 | 4 | 1702 | AAH02736.1 | |
| CR457270.1 | CAG33551.1 |
Target Gene Details
Entrez Gene ID: | 114971 |
Gene Name: | protein tyrosine phosphatase, mitochondrial 1 |
Gene Aliases: |
DUSP23, MOSP, PLIP, PNAS-129 |
Location: |
Chr.11:47565430-47573461 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPMT1 | NM_001143984.1 | 2 | 2026 | NP_001137456.1 |
| NM_175732.2 | 4 | 2109 | NP_783859.1 | |
| BC014048.1 | 3 | 1755 | AAH14048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss |
|
| nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain |
|
| nsv509405 | Chr.11:47571038 - 47663104 on Build GRCh38 | Insertion |
|
| nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map