Assay Details
Target Gene Details
Entrez Gene ID: | 10077 |
Gene Name: | tetraspanin 32 |
Gene Aliases: |
ART1, PHEMX, PHMX, TSSC6 |
Location: |
Chr.11:2299480-2318201 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN32 | NM_139022.2 | NP_620591.3 | ||
| XM_005252719.4 | XP_005252776.1 | |||
| XM_005252720.4 | XP_005252777.1 | |||
| XM_011519815.2 | XP_011518117.2 | |||
| XM_011519818.2 | XP_011518120.2 | |||
| XM_011519819.1 | XP_011518121.1 | |||
| XM_011519822.1 | XP_011518124.1 | |||
| XM_011519828.2 | XP_011518130.1 | |||
| XM_011519829.2 | XP_011518131.2 | |||
| XM_017017067.1 | XP_016872556.1 | |||
| AF125569.1 | 1 | 157 | AAD23580.1 | |
| AF176070.1 | 1 | 172 | AAG15840.1 | |
| AF176071.1 | AAG15841.1 | |||
| AK128812.1 | ||||
| AK298692.1 | ||||
| AK307638.1 | ||||
| AK310116.1 | ||||
| AY039001.1 | AAK84431.1 | |||
| AY303780.1 | AAP76185.1 | |||
| BC016693.1 | AAH16693.1 | |||
| BG506933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss |
|
| nsv553086 | Chr.11:2300156 - 2304351 on Build GRCh38 | Loss |
|
| nsv553081 | Chr.11:2291007 - 2312445 on Build GRCh38 | Loss |
|
| nsv467645 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
| nsv951285 | Chr.11:2256071 - 2310670 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map