Assay Details
Target Gene Details
Entrez Gene ID: | 57458 |
Gene Name: | transmembrane and coiled-coil domain family 3 |
Gene Aliases: |
- |
Location: |
Chr.12:94567124-94650562 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCC3 | NM_001301036.1 | 4 | 3100 | NP_001287965.1 |
| NM_020698.3 | 4 | 3081 | NP_065749.2 | |
| AB032971.1 | 4 | 3010 | ||
| AB073655.1 | 1 | 1576 | ||
| BC040535.1 | 4 | 3067 | AAH40535.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114247573] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map