Assay Details
Target Gene Details
Entrez Gene ID: | 23493 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif 2 |
Gene Aliases: |
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32 |
Location: |
Chr.6:125747592-125762243 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEY2 | NM_012259.2 | 5 | 2226 | NP_036391.1 |
| XM_017010627.1 | 5 | 3610 | XP_016866116.1 | |
| XM_017010628.1 | 6 | 3308 | XP_016866117.1 | |
| XM_017010629.1 | 5 | 2087 | XP_016866118.1 | |
| AJ249545.1 | 5 | 2085 | CAB56839.1 |
More Information
Set Membership: |
|

Genomic Map