Assay Details
Target Gene Details
Entrez Gene ID: | 3728 |
Gene Name: | junction plakoglobin |
Gene Aliases: |
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB |
Location: |
Chr.17:41754607-41786853 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JUP | NM_002230.2 | 14 | 2674 | NP_002221.1 |
| NM_021991.2 | NP_068831.1 | |||
| XM_006721873.2 | XP_006721936.1 | |||
| XM_006721874.2 | XP_006721937.1 | |||
| XM_006721875.1 | XP_006721938.1 | |||
| XM_006721878.1 | XP_006721941.1 | |||
| XM_011524753.2 | XP_011523055.1 | |||
| XM_011524755.1 | XP_011523057.1 | |||
| XM_011524756.1 | XP_011523058.1 | |||
| XM_011524757.2 | XP_011523059.1 | |||
| XM_011524758.1 | XP_011523060.1 | |||
| XM_017024588.1 | XP_016880077.1 | |||
| XM_017024589.1 | 14 | 2657 | XP_016880078.1 | |
| XM_017024590.1 | XP_016880079.1 | |||
| AK226051.1 | 14 | 2651 | ||
| BC000441.2 | AAH00441.2 | |||
| BC011865.2 | AAH11865.1 | |||
| M23410.1 | 14 | 2657 | AAA64895.1 | |
| Z68228.1 | 14 | 2660 | CAA92522.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73983656] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map