Assay Details
Target Gene Details
Entrez Gene ID: | 26018 |
Gene Name: | leucine rich repeats and immunoglobulin like domains 1 |
Gene Aliases: |
LIG-1, LIG1 |
Location: |
Chr.3:66378797-66501124 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIG1 | NM_015541.2 | 14 | 1934 | NP_056356.2 |
| XM_011533578.1 | 13 | 2391 | XP_011531880.1 | |
| XM_011533579.2 | 14 | 1734 | XP_011531881.1 | |
| XM_017006134.1 | 13 | 2460 | XP_016861623.1 | |
| XM_017006135.1 | 9 | 1257 | XP_016861624.1 | |
| XM_017006136.1 | 14 | 1798 | XP_016861625.1 | |
| AB050468.1 | 14 | 2439 | BAB40659.1 | |
| AF381545.1 | 14 | 1934 | AAK62357.1 | |
| AK126745.1 | 7 | 1302 | ||
| AK294984.1 | 6 | 936 | ||
| AK314103.1 | 14 | 2445 | ||
| AL117666.1 | 1 | 91 | CAB56036.1 | |
| AY730707.1 | 13 | 1951 | AAU44786.1 | |
| BC071561.1 | AAH71561.1 | |||
| BC140907.1 | 14 | 1940 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590546 | Chr.3:65487766 - 67232113 on Build GRCh38 | Gain |
|
| nsv516847 | Chr.3:66377858 - 66387532 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map