Assay Details
Target Gene Details
Entrez Gene ID: | 27177 |
Gene Name: | interleukin 36, beta |
Gene Aliases: |
FIL1, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1F8, IL-1H2, IL1-ETA, IL1F8, IL1H2 |
Location: |
Chr.2:113022091-113052867 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL36B | NM_014438.4 | NP_055253.2 | ||
| NM_173178.2 | NP_775270.1 | |||
| XM_011510962.1 | XP_011509264.1 | |||
| AF200494.1 | AAF69250.1 | |||
| AF201833.1 | AAF25213.1 | |||
| BC101831.1 | AAI01832.1 | |||
| BC101833.1 | AAI01834.1 |
More Information
Set Membership: |
|

Genomic Map