Assay Details
Target Gene Details
Entrez Gene ID: | 91010 |
Gene Name: | formin like 3 |
Gene Aliases: |
FHOD3, FRL2, WBP-3, WBP3 |
Location: |
Chr.12:49636499-49707414 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMNL3 | NM_175736.4 | 26 | 7027 | NP_783863.4 |
| NM_198900.2 | 25 | 6874 | NP_944489.2 | |
| XM_005269218.2 | 27 | 7121 | XP_005269275.1 | |
| XM_011538968.2 | 27 | 7611 | XP_011537270.1 | |
| XM_011538969.2 | 26 | 7495 | XP_011537271.1 | |
| XM_011538970.2 | 27 | 7610 | XP_011537272.1 | |
| XM_011538971.2 | 27 | 7186 | XP_011537273.1 | |
| XM_011538972.2 | 26 | 7456 | XP_011537274.1 | |
| XM_011538973.2 | 23 | 6601 | XP_011537275.2 | |
| XM_011538974.2 | 28 | 6943 | XP_011537276.1 | |
| AB095934.2 | 26 | 7027 | ||
| BC159100.1 | 26 | 7027 |
Target Gene Details
Entrez Gene ID: | 25766 |
Gene Name: | pre-mRNA processing factor 40 homolog B |
Gene Aliases: |
HYPC |
Location: |
Chr.12:49622717-49644669 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952177 | Chr.12:49641618 - 49653517 on Build GRCh38 | Deletion |
|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map