Assay Details
Target Gene Details
Entrez Gene ID: | 9863 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Gene Aliases: |
ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM |
Location: |
Chr.7:78017055-79453805 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGI2 | NM_001301128.1 | 21 | 5559 | NP_001288057.1 |
| NM_012301.3 | 22 | 5601 | NP_036433.2 | |
| XM_011516718.1 | 24 | 5958 | XP_011515020.1 | |
| XM_011516719.2 | 24 | 9375 | XP_011515021.1 | |
| XM_011516720.2 | 24 | 14365 | XP_011515022.1 | |
| XM_011516726.2 | 21 | 5031 | XP_011515028.1 | |
| XM_011516728.1 | 20 | 4567 | XP_011515030.1 | |
| XM_017012840.1 | 25 | 6087 | XP_016868329.1 | |
| XM_017012841.1 | 25 | 6084 | XP_016868330.1 | |
| XM_017012842.1 | 25 | 6081 | XP_016868331.1 | |
| XM_017012843.1 | 24 | 6045 | XP_016868332.1 | |
| XM_017012844.1 | 23 | 5961 | XP_016868333.1 | |
| XM_017012845.1 | 24 | 5952 | XP_016868334.1 | |
| XM_017012846.1 | 23 | 5916 | XP_016868335.1 | |
| XM_017012847.1 | 25 | 12324 | XP_016868336.1 | |
| XM_017012848.1 | 23 | 9243 | XP_016868337.1 | |
| XM_017012849.1 | 23 | 9237 | XP_016868338.1 | |
| AB014605.2 | 21 | 5540 | ||
| BC150277.1 | 21 | 5540 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607654 | Chr.7:77908647 - 78385012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map