Assay Details
Target Gene Details
Entrez Gene ID: | 6829 |
Gene Name: | SPT5 homolog, DSIF elongation factor subunit |
Gene Aliases: |
SPT5, SPT5H, Tat-CT1 |
Location: |
Chr.19:39445546-39476670 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT5H | NM_001111020.2 | 16 | 1458 | NP_001104490.1 |
| NM_001130824.1 | 16 | 1383 | NP_001124296.1 | |
| NM_001130825.1 | 15 | 1446 | NP_001124297.1 | |
| NM_001319990.1 | 16 | 1380 | NP_001306919.1 | |
| NM_001319991.1 | 15 | 1368 | NP_001306920.1 | |
| NM_003169.3 | 15 | 1522 | NP_003160.2 | |
| XM_017027174.1 | 11 | 980 | XP_016882663.1 | |
| AB000516.1 | 16 | 1432 | BAA24075.1 | |
| AB209257.1 | 14 | 1366 | BAD92494.1 | |
| AF040253.1 | 15 | 1486 | AAD02179.1 | |
| AK091476.1 | 3 | 1543 | ||
| AK296117.1 | 9 | 720 | ||
| AK302954.1 | 17 | 1356 | ||
| AK303480.1 | 15 | 1237 | ||
| BC024203.2 | 16 | 1315 | AAH24203.1 | |
| U56402.1 | 16 | 1327 | AAC51102.1 | |
| Y12790.1 | 16 | 1422 | CAA73326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523856 | Chr.19:39422918 - 39506832 on Build GRCh38 | Loss |
|
| nsv953573 | Chr.19:39444461 - 39522960 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map