Assay Details
Target Gene Details
Entrez Gene ID: | 3695 |
Gene Name: | integrin subunit beta 7 |
Gene Aliases: |
- |
Location: |
Chr.12:53191318-53207376 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB7 | NM_000889.2 | 3 | 281 | NP_000880.1 |
| NR_104181.1 | 2 | 158 | ||
| XM_005268851.3 | 3 | 238 | XP_005268908.1 | |
| XM_005268852.4 | 2 | 154 | XP_005268909.1 | |
| XM_006719376.3 | 3 | 350 | XP_006719439.1 | |
| AK298239.1 | ||||
| AK300960.1 | 2 | 132 | ||
| AK301523.1 | 2 | 102 | ||
| AK311703.1 | ||||
| BC015916.1 | 2 | 120 | AAH15916.1 | |
| DC296007.1 | 3 | 282 | ||
| M62880.1 | 2 | 162 | AAA59185.1 | |
| M68892.1 | 2 | 200 | AAA59184.1 | |
| S80335.1 | 2 | 200 | AAB21332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain |
|
| nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss |
|
| nsv470295 | Chr.12:53099603 - 53227927 on Build GRCh38 | Gain |
|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
| dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss |
|
| nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map