Assay Details
Target Gene Details
Entrez Gene ID: | 30817 |
Gene Name: | adhesion G protein-coupled receptor E2 |
Gene Aliases: |
CD312, EMR2, VBU |
Location: |
Chr.19:14732697-14778541 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE2 | NM_001271052.1 | 19 | 5582 | NP_001257981.1 |
| NM_013447.3 | 21 | 6041 | NP_038475.2 | |
| AK024426.1 | 11 | 4410 | BAB15716.1 | |
| AK024454.1 | 6 | 3623 | BAB15744.1 | |
| CN480316.1 | 1 | 443 | ||
| CR936819.1 | 15 | 5018 |
Target Gene Details
Entrez Gene ID: | 84449 |
Gene Name: | zinc finger protein 333 |
Gene Aliases: |
- |
Location: |
Chr.19:14689787-14733746 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF333 | NM_001300912.1 | 12 | 3001 | NP_001287841.1 |
| BC048107.1 | 12 | 2972 | AAH48107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578680 | Chr.19:14695350 - 14735523 on Build GRCh38 | Loss |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map