Assay Details
Target Gene Details
Entrez Gene ID: | 4968 |
Gene Name: | 8-oxoguanine DNA glycosylase |
Gene Aliases: |
HMMH, HOGG1, MUTM, OGH1 |
Location: |
Chr.3:9749944-9788246 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGG1 | XM_011533760.1 | XP_011532062.1 | ||
| XM_017006499.1 | XP_016861988.1 |
Target Gene Details
Entrez Gene ID: | 10474 |
Gene Name: | transcriptional adaptor 3 |
Gene Aliases: |
ADA3, NGG1, STAF54, TADA3L, hADA3 |
Location: |
Chr.3:9779964-9793011 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TADA3 | NM_001278270.1 | NP_001265199.1 | ||
| NM_006354.3 | NP_006345.1 | |||
| NM_133480.2 | NP_597814.1 | |||
| NR_103488.1 | ||||
| AF069733.1 | AAC39903.1 | |||
| AK000228.1 | ||||
| AK024982.1 | ||||
| AK292264.1 | ||||
| AL117487.1 | CAB55957.1 | |||
| BC009240.2 | AAH09240.1 | |||
| BC013433.1 | AAH13433.1 | |||
| BG477633.1 | ||||
| CR533543.1 | CAG38574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map