Assay Details
Target Gene Details
Entrez Gene ID: | 80006 |
Gene Name: | trafficking protein particle complex 13 |
Gene Aliases: |
C5orf44 |
Location: |
Chr.5:65624731-65666136 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC13 | NM_001093755.1 | NP_001087224.1 | ||
| NM_001093756.1 | NP_001087225.1 | |||
| NM_001243737.1 | NP_001230666.1 | |||
| NM_024941.3 | NP_079217.2 | |||
| NR_003545.1 | 2 | 845 | ||
| XM_011543652.2 | XP_011541954.1 | |||
| XM_017009890.1 | XP_016865379.1 | |||
| AK023673.1 | BAB14633.1 | |||
| AK130467.1 | 1 | 2263 | BAC85360.1 | |
| BC142994.1 | ||||
| BX641157.1 | CAE46070.1 | |||
| DA903408.1 | ||||
| DB145758.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894204 | Chr.5:65586109 - 65714937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map